CAS RN 73891-15-3
CAS RN 73891-15-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-(18-Crown-6)-2,3,11,12-tetracarboxylic Acid, TRC
CAS: 73891-15-3 Molecular Formula: C16H24O14 Molecular Weight (g/mol): 440.35 Synonym: (2S,3S,11S,12S)-1,4,7,10,13,16-Hexaoxacyclooctadecane-2,3,11,12-tetracarboxylic Acid,[2S-(2R*,3R*,11R*,12R*)]-1,4,7,10,13,16-Hexaoxacyclooctadecane-2,3,11,12-tetracarboxylic Acid IUPAC Name: (2S,3S,11S,12S)-1,4,7,10,13,16-hexaoxacyclooctadecane-2,3,11,12-tetracarboxylic acid SMILES: OC(=O)[C@H]1OCCOCCO[C@@H]([C@H](OCCOCCO[C@@H]1C(=O)O)C(=O)O)C(=O)O