accessibility menu, dialog, popup

UserName

CAS RN 7359-14-0

CH3OIOHO

CAS RN 7359-14-0

IUPAC Name: 2-hydroxy-5-iodo-3-methoxybenzaldehyde
Synonyms: 2-hydroxy-5-iodo-3-methoxybenzaldehyde
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: KVQIQUILJIOMHJ-UHFFFAOYSA-N
SMILES: COC1=CC(I)=CC(C=O)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results