accessibility menu, dialog, popup

UserName

HO(S)H2NSOOO

CAS RN 73463-39-5

IUPAC Name: (2S)-3-(cyclohexylazaniumyl)-2-hydroxypropane-1-sulfonate
Synonyms: (2S)-3-(cyclohexylammonio)-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol): 237.31
Molecular Formula: C9H19NO4S
InChi Key: INEWUCPYEUEQTN-VIFPVBQESA-N
SMILES: O[C@@H](C[NH2+]C1CCCCC1)CS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (8)
Quantity 
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (8)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)