CAS RN 7338-43-4
IUPAC Name:
(1S)-1-phenylpentan-1-ol
Synonyms:
(1S)-1-phenylpentan-1-ol
Molecular Weight (g/mol):
164.25
Molecular Formula:
C11H16O
InChi Key:
OVGORFFCBUIFIA-NSHDSACASA-N
SMILES:
CCCC[C@H](O)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Cyclohexyl-1-pentanol, 98%, Thermo Scientific™
CAS: 7338-43-4 Molecular Formula: C11H22O Molecular Weight (g/mol): 170.296 MDL Number: MFCD00045483 InChI Key: PKXSPCMDZCKLCI-UHFFFAOYSA-N Synonym: 1-cyclohexyl-1-pentanol,cyclohexylpentanol-1,1-cyclohexyl-pentan-1-ol,acmc-20aj12,n-butyl cyclohexyl carbinol,1-hydroxypentyl cyclohexane,1-cyclohexyl pentanol-1,pkxspcmdzcklci-uhfffaoysa,--1-cyclohexyl-1-pentanol PubChem CID: 145270 IUPAC Name: 1-cyclohexylpentan-1-ol SMILES: CCCCC(C1CCCCC1)O