accessibility menu, dialog, popup

UserName

CAS RN 7337-45-3

CH3CH3NH2H3CBH3

CAS RN 7337-45-3

IUPAC Name: boron(3+) 2-methylpropan-2-amine trihydride
Synonyms: borane tert-butylamine complex
Molecular Weight (g/mol): 86.97
Molecular Formula: C4H14BN
InChi Key: HCRXDJRJMXESSG-UHFFFAOYSA-N
SMILES: [H-].[H-].[H-].[B+3].CC(C)(C)N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)