accessibility menu, dialog, popup

UserName

CAS RN 7335-25-3

H3COOCl

CAS RN 7335-25-3

IUPAC Name: ethyl 2-chlorobenzoate
Synonyms: ethyl 2-chlorobenzoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: RETLCWPMLJPOTP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results