accessibility menu, dialog, popup

UserName

CAS RN 73282-11-8

SOONHO

CAS RN 73282-11-8

IUPAC Name: [1-(benzenesulfonyl)-1H-indol-2-yl]methanol
Synonyms: [1-(benzenesulfonyl)indol-2-yl]methanol
Molecular Weight (g/mol): 287.33
Molecular Formula: C15H13NO3S
InChi Key: LRYLVFIUTJMZBY-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results