accessibility menu, dialog, popup

UserName

CAS RN 7326-19-4

HO(R)HOO

CAS RN 7326-19-4

IUPAC Name: (2R)-2-hydroxy-3-phenylpropanoic acid
Synonyms: (R)-phenyllactate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: VOXXWSYKYCBWHO-MRVPVSSYSA-N
SMILES: O[C@H](CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)