accessibility menu, dialog, popup

UserName

CAS RN 7321-93-9

NClNH2Cl

CAS RN 7321-93-9

IUPAC Name: 4,6-dichloropyridin-3-amine
Synonyms: 4,6-dichloropyridin-3-amine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: FBGVTWONYOCYGA-UHFFFAOYSA-N
SMILES: NC1=CN=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results