accessibility menu, dialog, popup

UserName

CAS RN 73183-34-3

CH3H3COBOH3CH3CBOCH3CH3CH3H3CO

CAS RN 73183-34-3

IUPAC Name: 4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
Synonyms: bis(pinacolato)diboron
Molecular Weight (g/mol): 253.94
Molecular Formula: C12H24B2O4
InChi Key: IPWKHHSGDUIRAH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)