accessibility menu, dialog, popup

UserName

CAS RN 7312-11-0

H3COOSBr

CAS RN 7312-11-0

IUPAC Name: methyl 5-bromo-1-benzothiophene-2-carboxylate
Synonyms: methyl 5-bromo-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 271.13
Molecular Formula: C10H7BrO2S
InChi Key: XDYVZHUZZZKQOS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC(Br)=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results