accessibility menu, dialog, popup

UserName

CH3H3CSOONHOOHCH3CH3

CAS RN 730249-87-3

IUPAC Name: 4-(2,3,5,6-tetramethylbenzenesulfonamido)benzoic acid
Synonyms: 4-(2,3,5,6-tetramethylbenzenesulfonamido)benzoic acid
Molecular Weight (g/mol): 333.4
Molecular Formula: C17H19NO4S
InChi Key: GXRBZEBVKOIXDU-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(=C1C)S(=O)(=O)NC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results