accessibility menu, dialog, popup

UserName

CAS RN 72990-37-5

NOCl

CAS RN 72990-37-5

IUPAC Name: 3-chloropyridine-4-carbaldehyde
Synonyms: 3-chloropyridine-4-carbaldehyde
Molecular Weight (g/mol): 141.55
Molecular Formula: C6H4ClNO
InChi Key: ZVGDKOQPJCOCLI-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)