accessibility menu, dialog, popup

UserName

CAS RN 7287-19-6

H3CSNNHCH3CH3NNHH3CCH3N

CAS RN 7287-19-6

IUPAC Name: 6-(methylsulfanyl)-N2,N4-bis(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: selectin caswell no. 097 gesagarde 50 wp epa pesticide chemical code 080805 50, gesaguard Uvon selectin 50 gesaguard 50 prometryn prometrin
Molecular Weight (g/mol): 241.36
Molecular Formula: C10H19N5S
InChi Key: AAEVYOVXGOFMJO-UHFFFAOYSA-N
SMILES: CSC1=NC(NC(C)C)=NC(NC(C)C)=N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)