CAS RN 72835-25-7
IUPAC Name:
5-methylthiophene-2-carbonitrile
Synonyms:
5-methylthiophene-2-carbonitrile
Molecular Weight (g/mol):
123.17
Molecular Formula:
C6H5NS
InChi Key:
RBQRZWYCXAXPIN-UHFFFAOYSA-N
SMILES:
CC1=CC=C(S1)C#N
1
–
2
of
2
results
2
5-Methylthiophene-2-carbonitrile, 97%, Thermo Scientific™
CAS: 72835-25-7 Molecular Formula: C6H5NS Molecular Weight (g/mol): 123.18 InChI Key: RBQRZWYCXAXPIN-UHFFFAOYSA-N PubChem CID: 312733 IUPAC Name: 5-methylthiophene-2-carbonitrile SMILES: CC1=CC=C(S1)C#N