CAS RN 72580-53-1
IUPAC Name:
pyrrolidine-3-carboxylic acid
Synonyms:
pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol):
115.13
Molecular Formula:
C5H9NO2
InChi Key:
JAEIBKXSIXOLOL-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCNC1
1
–
1
of
1
results
1
L-beta-Proline, 98+%
CAS: 72580-53-1 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.132 MDL Number: MFCD06738463 InChI Key: JAEIBKXSIXOLOL-BYPYZUCNSA-N Synonym: s-pyrrolidine-3-carboxylic acid,s-+-pyrrolidine-3-carboxylic acid,l-beta-proline,3s-3-pyrrolidinecarboxylic acid,3s-pyrrolidine-3-carboxylic acid,3-pyrrolidinecarboxylic acid, 3s,s-pyrrolidine-3-carboxylicacid,s-beta-proline,s-,a-proline,pubchem11126 PubChem CID: 1501969 IUPAC Name: (3S)-pyrrolidine-3-carboxylic acid SMILES: C1CNCC1C(=O)O