accessibility menu, dialog, popup

UserName

CAS RN 7253-07-8

BrP

CAS RN 7253-07-8

IUPAC Name: (9H-fluoren-9-yl)triphenylphosphanium bromide
Synonyms: 9H-fluoren-9-yltriphenylphosphanium bromide
Molecular Weight (g/mol): 507.41
Molecular Formula: C31H24BrP
InChi Key: BHNPKGAMAZKCCJ-UHFFFAOYSA-M
SMILES: [Br-].C1=CC=C(C=C1)[P+](C1C2=CC=CC=C2C2=CC=CC=C12)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)