accessibility menu, dialog, popup

UserName

CAS RN 72432-10-1

IUPAC Name: 1-(4-methoxybenzoyl)pyrrolidin-2-one
Synonyms: aniracetam 茴拉西坦 ampamet sarpul aniracetamun 1-P-anisoyl-2-pyrrolidinone draganon
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: ZXNRTKGTQJPIJK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N1CCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)