CAS RN 7236-57-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Thiothymidine, TRC
CAS: 7236-57-9 Molecular Formula: C10H14N2O4S Molecular Weight (g/mol): 258.29 IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-4-sulfanylidenepyrimidin-2-one SMILES: CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=S