accessibility menu, dialog, popup

UserName

CAS RN 723281-72-9

NBrClNOO

CAS RN 723281-72-9

IUPAC Name: 6-bromo-4-chloro-3-nitroquinoline
Synonyms: 6-bromo-4-chloro-3-nitroquinoline
Molecular Weight (g/mol): 287.5
Molecular Formula: C9H4BrClN2O2
InChi Key: KHKPVEMMXJCWGP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C2=CC(Br)=CC=C2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results