accessibility menu, dialog, popup

UserName

ClOO

CAS RN 7228-38-8

IUPAC Name: 5-chloro-2H-1,3-benzodioxole
Synonyms: 5-chloro-2H-1,3-benzodioxole
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: ODQPZHOXLYATLC-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OCOC2=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results