accessibility menu, dialog, popup

UserName

CAS RN 7223-38-3

CH3NH3CHC

CAS RN 7223-38-3

IUPAC Name: dimethyl(prop-2-yn-1-yl)amine
Synonyms: dimethyl(prop-2-yn-1-yl)amine
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: ILBIXZPOMJFOJP-UHFFFAOYSA-N
SMILES: CN(C)CC#C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)