CAS RN 721401-53-2
CAS RN 721401-53-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5-Chloro-N-[(2R)-2-hydroxy-3-[[4-(3-oxomorpholin-4-yl)phenyl]amino]propyl]thiophene-2-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Decarbonyl Rivaroxaban, TRC
CAS: 721401-53-2 Molecular Formula: C18 H20 Cl N3 O4 S Molecular Weight (g/mol): 409.89 Synonym: 5-Chloro-N-[(2R)-2-hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-2-thiophencarboxamide,5-Chloro-N-[(2R)-2-hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-2-thiophenecarboxamide IUPAC Name: 5-chloro-N-[(2R)-2-hydroxy-3-[4-(3-oxomorpholin-4-yl)anilino]propyl]thiophene-2-carboxamide SMILES: O[C@@H](CNC(=O)c1ccc(Cl)s1)CNc2ccc(cc2)N3CCOCC3=O