CAS RN 721-90-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenylalanylglycine, TRC
CAS: 721-90-4 Molecular Formula: C11H14N2O3 Molecular Weight (g/mol): 222.24 Synonym: L-Phenylalanylglycine,105: PN: US20130123467 SEQID: 128 Claimed Protein,10: PN: WO2014063098 TABLE: 5 Claimed Protein,2-[(2S)-2-Amino-3-phenylpropanamido]acetic Acid,2-[[(2S)-2-Amino-3-phenylpropanoyl]amino]acetic Acid,28: PN: WO2016049036 SEQID: 69 Claimed Protein,3: PN: RU2506269 PAGE: 14 Claimed Sequence,NSC 89182 IUPAC Name: 2-[[(2S)-2-amino-3-phenylpropanoyl]amino]acetic acid SMILES: N[C@@H](Cc1ccccc1)C(=O)NCC(=O)O