accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 720-75-2

IUPAC Name: methyl [1,1'-biphenyl]-4-carboxylate
Synonyms: methyl [1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: GATUGNVDXMYTJX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results