accessibility menu, dialog, popup

UserName

CAS RN 71957-08-9

CoHO(S)OH(S)OH(R)OH(S)OHOHO

CAS RN 71957-08-9

IUPAC Name: λ²-cobalt(2+) (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanoic acid
Synonyms: λ²-cobalt(2+) (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanoic acid
Molecular Weight (g/mol): 255.09
Molecular Formula: C6H12CoO7
InChi Key: ADBDSOAUIVHRFK-OSQBQZLYSA-N
SMILES: [Co++].OC[C@H](O)[C@H](O)[C@@H](O)[C@H](O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results