accessibility menu, dialog, popup

UserName

CAS RN 718596-77-1

NOHOH2N

CAS RN 718596-77-1

IUPAC Name: 3-amino-3-(4-cyanophenyl)propanoic acid
Synonyms: 3-amino-3-(4-cyanophenyl)propanoic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: JFPLLJRLBHIJPS-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)