accessibility menu, dialog, popup

UserName

CAS RN 7184-60-3

CH3CH3H3COHOOOHH3CHOON(E)(E)

CAS RN 7184-60-3

IUPAC Name: 2-[(4E,6E)-7-cyano-8,16-dihydroxy-9,11,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6-dien-2-yl]cyclopentane-1-carboxylic acid
Synonyms: 2-[(4E,6E)-7-cyano-8,16-dihydroxy-9,11,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6-dien-2-yl]cyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 489.65
Molecular Formula: C28H43NO6
InChi Key: OJCKRNPLOZHAOU-SLNPHPKONA-N
SMILES: CC1CC(C)CC(C)C(O)\C(=C\C=C\CC(OC(=O)CC(O)C(C)C1)C1CCCC1C(O)=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results