missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borrelidin, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Antibiotic and antiangiogenic. Selectively inhibits threonyl-tRNA synthetase (ThrRS). Exhibits antiangiogenic activity in a mouse model of tumor angiogenesis. Induces apoptosis in capillary tube-forming endothelial cells; disrupts capillary tubes and inhibits their formation (IC50 = 0.8 nM in rat aorta). Induces the unfolded protein response (UPR) and apoptosis in oral cancer cells.
Specifications
Specifications
| CAS | 7184-60-3 |
| Molecular Formula | C28H43NO6 |
| Quantity | 1 mg |
| Synonym | treponemycin |
| InChI Key | OJCKRNPLOZHAOU-MJQQLCDASA-N |
| SMILES | CC1CC(CC(C(C(=CC=CCC(OC(=O)CC(C(C1)C)O)C2CCCC2C(=O)O)C#N)O)C)C |
| IUPAC Name | (1S,2S)-2-[(2R,4Z,6E,8S,9R,13R,15R,16R)-7-cyano-8,16-dihydroxy-9,11,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6-dien-2-yl]cyclopentane-1-carboxylic acid |
| Molecular Weight (g/mol) | 489.653 |
| PubChem CID | 133655849 |
| Formula Weight | 489.64 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction