CAS RN 71776-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Dimethylbutylamine Hydrochloride, TRC
CAS: 71776-70-0 Molecular Formula: C6H16ClN Molecular Weight (g/mol): 137.65 Synonym: (±)-1,3-Dimethylbutylamine Hydrochloride,(±)-4-Methyl-2-Pentanamine Hydrochloride,1,3-Dimethylbutanamine Hydrochloride,2-Amino-4-methylpentane Hydrochloride,4-Methyl-2-aminopentane Hydrochloride,4-Methyl-2-pentanamine Hydrochloride,DL-1,3-Dimethylbutylamine Hydrochloride,NSC 48080 Hydrochloride IUPAC Name: 4-methylpentan-2-amine;hydrochloride SMILES: CC(C)CC(C)N.Cl