accessibility menu, dialog, popup

UserName

CH3OFNH2F

CAS RN 717094-51-4

IUPAC Name: 1-(2,4-difluoro-3-methoxyphenyl)methanamine
Synonyms: 1-(2,4-difluoro-3-methoxyphenyl)methanamine
Molecular Weight (g/mol): 173.16
Molecular Formula: C8H9F2NO
InChi Key: AAZMLHVLBRWOGC-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(CN)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results