accessibility menu, dialog, popup

UserName

CAS RN 71574-33-9

CH3SNH2NCH3HCl

CAS RN 71574-33-9

IUPAC Name: dimethyl-1,3-thiazol-2-amine hydrochloride
Synonyms: dimethyl-1,3-thiazol-2-amine hydrochloride
Molecular Weight (g/mol): 164.65
Molecular Formula: C5H9ClN2S
InChi Key: XYTUTNQRQLAZLK-UHFFFAOYSA-N
SMILES: Cl.CC1=C(C)N=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results