accessibility menu, dialog, popup

UserName

CH3ONHCH3Cl

CAS RN 7149-79-3

IUPAC Name: N-(3-chloro-4-methylphenyl)acetamide
Synonyms: 3-chloro-4-methylacetanilide
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: IGLVCWPPISIXPR-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results