accessibility menu, dialog, popup

UserName

H3CBrClO

CAS RN 7148-74-5

IUPAC Name: 2-bromopropanoyl chloride
Synonyms: 2-bromo-propionyl chloride
Molecular Weight (g/mol): 171.42
Molecular Formula: C3H4BrClO
InChi Key: OZGMODDEIHYPRY-UHFFFAOYNA-N
SMILES: CC(Br)C(Cl)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)