accessibility menu, dialog, popup

UserName

H3COONCH3CH3

CAS RN 7148-06-3

IUPAC Name: methyl 2-(dimethylamino)acetate
Synonyms: methyl 2-(dimethylamino)acetate
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: LRZFEBJUJIQVDQ-UHFFFAOYSA-N
SMILES: COC(=O)CN(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)