accessibility menu, dialog, popup

UserName

H3CSN

CAS RN 71037-08-6

IUPAC Name: 2-(butylsulfanyl)acetonitrile
Synonyms: 2-(butylsulfanyl)acetonitrile
Molecular Weight (g/mol): 129.22
Molecular Formula: C6H11NS
InChi Key: LRMUBLYDYGZCDR-UHFFFAOYSA-N
SMILES: CCCCSCC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)