accessibility menu, dialog, popup

UserName

NH2NOHN

CAS RN 71-30-7

IUPAC Name: 6-amino-1,2-dihydropyrimidin-2-one
Synonyms: 2(1H)-pyrimidinone, 6-amino-
Molecular Weight (g/mol): 111.1
Molecular Formula: C4H5N3O
InChi Key: OPTASPLRGRRNAP-UHFFFAOYSA-N
SMILES: NC1=CC=NC(=O)N1
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (5)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)