accessibility menu, dialog, popup

UserName

CAS RN 709-63-7

H3COFFF

CAS RN 709-63-7

IUPAC Name: 1-[4-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: p-(trifluoromethyl)benzoyl
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: HHAISVSEJFEWBZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)