accessibility menu, dialog, popup

UserName

CAS RN 7064-38-2

H3CNOCH3NOOCH3SSNOHOON(Z)

CAS RN 7064-38-2

IUPAC Name: 6-[(5Z)-5-({5-cyano-2-[3-(ethoxycarbonyl)piperidin-1-yl]-4-methyl-6-oxo-1-propyl-1,6-dihydropyridin-3-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]hexanoic acid
Synonyms: 6-[(5Z)-5-({5-cyano-2-[3-(ethoxycarbonyl)piperidin-1-yl]-4-methyl-6-oxo-1-propylpyridin-3-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]hexanoic acid
Molecular Weight (g/mol): 588.74
Molecular Formula: C28H36N4O6S2
InChi Key: GDARZPKSXSXRDC-JCMHNJIXNA-N
SMILES: CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCCCCC(O)=O)C2=O)=C1N1CCCC(C1)C(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results