accessibility menu, dialog, popup

UserName

CAS RN 705254-34-8

BOOCH3FFFK

CAS RN 705254-34-8

IUPAC Name: potassium trifluoro[4-(methoxycarbonyl)phenyl]boranuide
Synonyms: potassium trifluoro[4-(methoxycarbonyl)phenyl]boranuide
Molecular Weight (g/mol): 242.05
Molecular Formula: C8H7BF3KO2
InChi Key: IQYSKWRPRJKKAG-UHFFFAOYSA-N
SMILES: [K+].COC(=O)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results