accessibility menu, dialog, popup

UserName

CH3OONOOH3COOCH3

CAS RN 70384-51-9

IUPAC Name: 8-[2-(2-methoxyethoxy)ethyl]-2,5,11,14-tetraoxa-8-azapentadecane
Synonyms: 8-[2-(2-methoxyethoxy)ethyl]-2,5,11,14-tetraoxa-8-azapentadecane
Molecular Weight (g/mol): 323.43
Molecular Formula: C15H33NO6
InChi Key: XGLVDUUYFKXKPL-UHFFFAOYSA-N
SMILES: COCCOCCN(CCOCCOC)CCOCCOC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)