accessibility menu, dialog, popup

UserName

CAS RN 70380-70-0

IUPAC Name: 5-(thiophen-2-yl)-1,3-oxazole
Synonyms: 5-(thiophen-2-yl)-1,3-oxazole
Molecular Weight (g/mol): 151.18
Molecular Formula: C7H5NOS
InChi Key: VGBKCFRTMPSJLX-UHFFFAOYSA-N
SMILES: O1C=NC=C1C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results