accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 7035-03-2

IUPAC Name: 2-(2-methoxyphenyl)acetonitrile
Synonyms: 2-(2-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: DWJKILXTMUGXOU-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1CC#N
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results