accessibility menu, dialog, popup

UserName

CAS RN 703-80-0

H3COHN

CAS RN 703-80-0

IUPAC Name: 1-(1H-indol-3-yl)ethan-1-one
Synonyms: 3-acetyl indole
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: VUIMBZIZZFSQEE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)