accessibility menu, dialog, popup

UserName

CAS RN 7028-57-1

IUPAC Name: 2-methyl-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 2-methyl-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: MCEOSIYJRZMQNS-UHFFFAOYNA-N
SMILES: CC1SC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results