accessibility menu, dialog, popup

UserName

ClCl

CAS RN 7012-16-0

IUPAC Name: 2,7-dichloro-9H-fluorene
Synonyms: 2,7-dichloro-9H-fluorene
Molecular Weight (g/mol): 235.11
Molecular Formula: C13H8Cl2
InChi Key: SDPURBHAHVFTGX-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(CC3=CC(Cl)=CC=C23)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)