accessibility menu, dialog, popup

UserName

CAS RN 70028-88-5

IUPAC Name: phenyl(3,4,5-trifluorophenyl)methanone
Synonyms: phenyl(3,4,5-trifluorophenyl)methanone
Molecular Weight (g/mol): 236.19
Molecular Formula: C13H7F3O
InChi Key: BTQOMZVQASIULU-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1F)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results