accessibility menu, dialog, popup

UserName

FNCOFFF

CAS RN 69922-27-6

IUPAC Name: 1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene
Synonyms: 1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: NAIKHCBDZGSGHH-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1N=C=O)C(F)(F)F
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results