accessibility menu, dialog, popup

UserName

NNHNHBrHBr

CAS RN 699020-93-4

IUPAC Name: 4-[(1H-imidazol-5-yl)methyl]pyridine dihydrobromide
Synonyms: immethridine
Molecular Weight (g/mol): 321.02
Molecular Formula: C9H11Br2N3
InChi Key: CYNKWHIKNDIVDR-UHFFFAOYSA-N
SMILES: Br.Br.C(C1=CN=CN1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results