accessibility menu, dialog, popup

UserName

OOHNHOO

CAS RN 69901-75-3

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-2-cyclohexylacetic acid
Synonyms: {[(benzyloxy)carbonyl]amino}(cyclohexyl)acetic acid
Molecular Weight (g/mol): 291.35
Molecular Formula: C16H21NO4
InChi Key: CUSYTUPJAYLNFQ-UHFFFAOYNA-N
SMILES: OC(=O)C(NC(=O)OCC1=CC=CC=C1)C1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)